Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Oxa-3-aza-bicyclo[3.2.1]octane 98%

Supplier: APOLLO SCIENTIFIC

OR923369-1G OR923369-250MG
77411-842EA 296.75 USD InStock
77411-842 77411-844
8-Oxa-3-aza-bicyclo[3.2.1]octane 98%
8-Oxa-3-aza-bicyclo[3.2.1]octane


Formula: C₆H₁₁NO
Boiling Pt: 176…178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09836264
CAS Number: 280-13-7
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty