Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromo(difluoro)acetic anhydride

Supplier: APOLLO SCIENTIFIC
Danger

PC53049-100G PC53049-25G PC53049-5G
77416-608EA 365.76 USD InStock
77416-608 77416-610 77416-612
Bromo(difluoro)acetic anhydride
Bromo(difluoro)acetic anhydride


Formula: C₄Br₂F₄O₃
Boiling Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD24842567
CAS Number: 7601-98-1
UN: 3265
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty