Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-fluoro-2-methylphenol

Supplier: APOLLO SCIENTIFIC
Warning

PC104119-5G PC104119-1G PC104119-250MG
77412-854EA 619.44 USD InStock
77412-854 77412-850 77412-852
3-Bromo-5-fluoro-2-methylphenol
3-Bromo-5-fluoro-2-methylphenol


Formula: C₇H₆BrFO
MDL Number: MFCD28527221
CAS Number: 1784522-73-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty