Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (APC/iFluor® 700)

Supplier: AAT BIOQUEST INC
104411F0 104411F1 104411F2
77022-674EA 210.7 USD
77022-674 77022-920 77021-972
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (APC/iFluor® 700)
Antibodies
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Precaución: For research use only.

Type: Secondary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: APC/iFluor® 700
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen Symbol CD44
Environmentally Preferable
Antigen Name CD44 antigen
Antibody Type Secondary
Clonality Monoclonal
Conjugation APC (Allophycocyanin)/iFluor® 700
Clone IM7
Reactivity Human
Host Rat
Isotype
Purification Affinity purification
Storage Temperature Refrigerated (2...8 °C); Minimize light exposure
Shipping Temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR