Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl trifluoroacetate ≥99.0%

Supplier: Decon Laboratories Ltd

T0432-500G T0432-25G
TCT0432-500GEA 339 USD InStock
TCT0432-500G TCT0432-25G
Ethyl trifluoroacetate ≥99.0%
Ethyl trifluoroacetate
CAS Number: 383-63-1
MDL Number: MFCD00000419
Molecular Formula: C4H5F3O2
Molecular Weight: 142.08
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 62
Flash Point (°C): -7
Specific Gravity (20/20): 1.19

Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Trifluoroacetic Acid Ethyl Ester


Formula: C₄H₅F₃O₂
Boiling Pt: 62 °C
Melting Pt: –78 °C
Density: 1.19 (20 °C)
Flash Pt: –7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000419
CAS Number: 383-63-1
EINECS: 206-851-6
UN: 2924
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=488558da2b6ced993030ce81714bf616df6e5e78&vid=488558da2b6ced993030ce81714bf616df6e5e78&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty