Permanent Adhesive Poly Bags with ¹/₄" Vent Hole, Associated Bag
Supplier: Associated Bag
221-25 221-35
47751-434CS 281.69 USD
47751-434 47751-436
Permanent Adhesive Poly Bags with ¹/₄" Vent Hole, Associated Bag
Bags Storage Bags
Peel-and-seal adhesive strip forms a permanent, tamper-evident seal that must be cut or torn to open.
Bottom ¹/₄" vent hole allows air to circulate and moisture to escape.