Brinkmann® CH-500 Column Heater, Metrohm
Supplier: Metrohm USA
022852001 022852035
21160-000EA 5873.12 USD InStock
21160-000 21160-004
Brinkmann® CH-500 Column Heater, Metrohm
Heaters Column Heaters
This one-piece HPLC heating system offers temperature accuracy and flexibility with a built-in temperature controller that has a range from ambient to 150 °C
The conductivity of the heating block allows set and column temperature to be equal. Modular heating blocks are included to fit 6, 10, and 13mm column widths. Unit saves space and can be mounted vertically.
Certificaciones: CE marked, UL listed, and CSA approved.
Información para pedidos: Two sets of 4" blocks and one set of 2" blocks are included.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty