Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[Bis(3,5-trifluoromethylphenylphosphino)-3,6-dimethoxy]- 2',6'-dimethylamino-1,1'-biphenyl ≥98%

Supplier: AMBEED, INC
Warning

A705557-5G A705557-100MG
77747-410EA 2198 USD InStock
77747-410 77747-409
2-[Bis(3,5-trifluoromethylphenylphosphino)-3,6-dimethoxy]- 2',6'-dimethylamino-1,1'-biphenyl ≥98%
2-[Bis(3,5-trifluoromethylphenylphosphino)-3,6-dimethoxy]- 2',6'-dimethylamino-1,1'-biphenyl
Precaución: for Research Use Only


Formula: C₃₄H₂₉F₁₂N₂O₂P
MDL Number: MFCD29905024
CAS Number: 1810068-30-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty