Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyanine 5 monosuccinimidyl ester 95%

Supplier: AMBEED, INC
Warning

A162586-50MG A162586-100MG
77567-374EA 311 USD InStock
77567-374 77611-584
Cyanine 5 monosuccinimidyl ester 95%
Cyanine 5 monosuccinimidyl ester
Precaución: For research use only.


Formula: C₃₇H₄₃N₃O₁₀S₂
MDL Number: MFCD29472578
CAS Number: 146368-14-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty