Amberlyst® 15 (H) H+≥4,7 mmol/g, dry
Supplier: AMBEED, INC
|
Warning
|
A920554-1000G
A920554-500G
77569-642EA
113
USD
77569-642
77595-484
Amberlyst® 15 (H) H+≥4,7 mmol/g, dry
Amberlyst® 15 (H)
Precaución: For research use only.
Formula:
(C₁₀H₁₀·C₈H₈O₃S)x |
MDL Number:
MFCD00145841 CAS Number: 39389-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...