Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(1,1-Dimethyl-2-(5-(1,1,3-trimethyl-1H-benzo[e]indol-3-ium-2-yl)penta-2,4-dien-1-ylidene)-1,2-dihydro-3H-benzo[e]indol-3-yl)hexanoate ≥98%

Supplier: AMBEED, INC

A1219644-10MG A1219644-5MG
77621-008EA 154.5 USD InStock
77621-008 77621-010
6-(1,1-Dimethyl-2-(5-(1,1,3-trimethyl-1H-benzo[e]indol-3-ium-2-yl)penta-2,4-dien-1-ylidene)-1,2-dihydro-3H-benzo[e]indol-3-yl)hexanoate ≥98%
6-(1,1-Dimethyl-2-(5-(1,1,3-trimethyl-1H-benzo[e]indol-3-ium-2-yl)penta-2,4-dien-1-ylidene)-1,2-dihydro-3H-benzo[e]indol-3-yl)hexanoate
Precaución: for Research Use Only


Formula: C₄₀H₄₂N₂O₂
MDL Number: MFCD28385497
CAS Number: 1449612-07-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty