Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Patton-Reeders reagent (Calconcarboxylic acid) indicator

Supplier: AMBEED, INC
Warning

A1402789-25G A1402789-1000G A1402789-500G
77597-148EA 26.65 USD InStock
77597-148 77622-044 77622-046
Patton-Reeders reagent (Calconcarboxylic acid) indicator
Patton-Reeders reagent (Calconcarboxylic acid)
Precaución: for Research Use Only


Formula: C₂₁H₁₄N₂O₇S
MDL Number: MFCD00004078
CAS Number: 3737-95-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty