Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Chloro-1H-indole-4-carboxylic acid ≥98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A216402-100MG A216402-250MG
77683-620EA 21 USD InStock
77683-620 77683-621
7-Chloro-1H-indole-4-carboxylic acid ≥98%, Ambeed.Inc
7-Chloro-1H-indole-4-carboxylic acid
Precaución: For research use only.


Formula: C₉H₆ClNO₂
MDL Number: MFCD11845605
CAS Number: 588688-45-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty