Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Monomethyl sebacate ≥98%

Supplier: AMBEED, INC
Warning

A248963-25G A248963-100G
77601-004EA 296.23 USD InStock
77601-004 77668-032
Monomethyl sebacate ≥98%
Monomethyl sebacate
Precaución: for Research Use Only


Formula: C₁₁H₂₀O₄
MDL Number: MFCD00004439
CAS Number: 818-88-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty