Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-4-(Aminomethyl)cyclohexanecarboxylic acid ≥98%

Supplier: AMBEED, INC
Warning

A251486-100MG A251486-1G
77746-399EA 140 USD InStock
77746-399 77746-400
cis-4-(Aminomethyl)cyclohexanecarboxylic acid ≥98%
cis-4-(Aminomethyl)cyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₈H₁₅NO₂
MDL Number: MFCD19706018
CAS Number: 1197-17-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty