Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vitamin B12a hydrochloride ≥98%

Supplier: AMBEED, INC
Warning

A352993-100MG A352993-1G
77668-640EA 41 USD InStock
77668-640 77668-642
Vitamin B12a hydrochloride ≥98%
Vitamin B12a hydrochloride
Precaución: for Research Use Only


Formula: C₆₂H₈₉CoN₁₃O₁₅P·HCl
MW: 1346.35 g/mol
MDL Number: MFCD00167463
CAS Number: 58288-50-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty