Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

UNC0321 tri(trifluoroacetate salt) ≥98%

Supplier: AMBEED, INC
Warning

A354382-10MG A354382-1MG
77746-693EA 198 USD InStock
77746-693 77746-694
UNC0321 tri(trifluoroacetate salt) ≥98%
UNC0321 tri(trifluoroacetate salt)
Precaución: for Research Use Only


Formula: C₂₇H₄₅N₇O₃
MDL Number: MFCD18251565
CAS Number: 1238673-32-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty