Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3S,3aR,4S,4aR,7aR,8R,9aR)-3,4a,8-Trimethyl-2,5-dioxo-2,3,3a,4,4a,5,7a,8,9,9a-decahydroazuleno[6,5-b]furan-4-yl (Z)-2-methylbut-2-enoate ≥98%

Supplier: AMBEED, INC
Warning

A426347-10MG A426347-5MG
77686-169EA 124 USD InStock
77686-169 77686-170
(3S,3aR,4S,4aR,7aR,8R,9aR)-3,4a,8-Trimethyl-2,5-dioxo-2,3,3a,4,4a,5,7a,8,9,9a-decahydroazuleno[6,5-b]furan-4-yl (Z)-2-methylbut-2-enoate ≥98%
(3S,3aR,4S,4aR,7aR,8R,9aR)-3,4a,8-Trimethyl-2,5-dioxo-2,3,3a,4,4a,5,7a,8,9,9a-decahydroazuleno[6,5-b]furan-4-yl (Z)-2-methylbut-2-enoate
Precaución: for Research Use Only


Formula: C₂₀H₂₆O₅
MDL Number: MFCD04034623
CAS Number: 16503-32-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty