Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

NVP-AEW541 ≥99%

Supplier: AMBEED, INC
Warning

A462988-1MG A462988-5MG A462988-10MG
77746-947EA 30 USD InStock
77746-947 77746-948 77746-946
NVP-AEW541 ≥99%
NVP-AEW541
Precaución: for Research Use Only


Formula: C₂₇H₂₉N₅O
MDL Number: MFCD22741515
CAS Number: 475489-16-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty