Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

racemic-2-Di-t-butylphosphino-1,1'-binaphthyl ≥98%

Supplier: AMBEED, INC
Warning

A473583-10G A473583-250MG A473583-25G A473583-5G
77669-126EA 1415 USD InStock
77669-126 77669-128 77669-130 77669-132
racemic-2-Di-t-butylphosphino-1,1'-binaphthyl ≥98%
racemic-2-Di-t-butylphosphino-1,1'-binaphthyl
Precaución: for Research Use Only


Formula: C₂₈H₃₁P
MDL Number: MFCD03094576
CAS Number: 255836-67-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty