Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ginsenoside Rb1 ≥98%

Supplier: AMBEED, INC
Warning

A604407-10MG A604407-5MG A604407-1MG
77617-528EA 14.42 USD InStock
77617-528 77617-530 77749-242
Ginsenoside Rb1 ≥98%
Ginsenoside Rb1
Precaución: for Research Use Only


Formula: C₅₄H₉₂O₂₃
MDL Number: MFCD00133367
CAS Number: 41753-43-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty