Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

EDTA tripotassium salt ≥98%

Supplier: AMBEED, INC
Warning

A632440-100G A632440-500G
77669-712EA 51 USD InStock
77669-712 77669-714
EDTA tripotassium salt ≥98%
EDTA tripotassium salt
Precaución: for Research Use Only


Formula: C₁₀H₁₃K₃N₂O₈
MDL Number: MFCD00013304
CAS Number: 17572-97-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty