Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt ≥98%

Supplier: AMBEED, INC
Warning

A909884-1G A909884-250MG A909884-100MG
77747-674EA 466 USD InStock
77747-674 77747-675 77747-673
1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt ≥98%
1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt
Precaución: for Research Use Only


Formula: C₄₂H₇₈NaO₁₀P
MDL Number: MFCD01310528
CAS Number: 67254-28-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty