Nalgene® PMP Jars, Straight-Sided, Thermo Scientific
Supplier: Thermo Fisher Scientific
2117-0060 2117-0125 2117-0250 2117-0500 2117-1000
16129-354CS 359.04 USD
16129-354 16129-356 16129-378 16129-390 16129-414
Nalgene® PMP Jars, Straight-Sided, Thermo Scientific
Jars General Use Jars
Transparent, autoclavable, with straight sides
Top is recessed to allow stacking. Jars of 125mL and 250mL sizes have standard mason jar threads. Ideal for museum jars — formaldehyde loss only about 2% per year. Remove closure before autoclaving, or set closure on top of jar at an angle without engaging the threads.
Suitable for food and beverage use. Meets requirements of CFR21, Part 177.1520 section of Food Additives Amendment of Federal Food and Drug Act.
Polypropylene white screw closures are linerless, noncontaminating, and autoclavable.