Funnels, Stainless Steel
Supplier: SAMPLING SYSTEMS LTD
A244-100 A244-120 A244-200 A244-250 A244-310 A244-50
77577-764EA 51.32 USD
77577-764 77577-766 77577-768 77577-770 77577-772 77577-774
Funnels, Stainless Steel
Funnels Filtration Funnels Filter Funnels
These stainless steel funnels are ideal for use with many type of liquids. Made from durable 304 grade stainless steel.
  • Crevice free construction
  • Fluted spout to aid product flow
  • Excellent chemical and temperature resistance