RecombiMAb Human IgG4 S228P L235E P329G (SPLEPG) Isotype control Anti-Hen Egg Lysozyme

Supplier: Bio X Cell
CP148-1MG CP148-5MG
77580-232EA 238.36 USD
77580-232 77580-238
RecombiMAb Human IgG4 S228P L235E P329G (SPLEPG) Isotype control Anti-Hen Egg Lysozyme
Antibodies
Type: Control
Antigen: IgG4
Clonality: Recombinant
Clone: N/A-CP148
Conjugation:
Epitope:
Host:
Isotype: IgG4
Reactivity: Hen
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR