Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-((6,7-Dimethoxyquinazolin-4-yl)oxy)-N-(4-((4-ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)-4-methylbenzamide 97%

Supplier: AMBEED, INC
Warning

A929460-10MG A929460-5MG
77747-691EA 105 USD InStock
77747-691 77747-692
3-((6,7-Dimethoxyquinazolin-4-yl)oxy)-N-(4-((4-ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)-4-methylbenzamide 97%
3-((6,7-Dimethoxyquinazolin-4-yl)oxy)-N-(4-((4-ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)-4-methylbenzamide
Precaución: for Research Use Only


Formula: C₃₂H₃₄F₃N₅O₄
MDL Number: MFCD29991221
CAS Number: 1315330-17-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty