Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Phe(4-CN)-OH ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

B4877-1G B4877-5G
TCB4877-1GEA 53 USD InStock
TCB4877-1G TCB4877-5G
Boc-Phe(4-CN)-OH ≥98.0% (by HPLC, titration analysis)
Boc-Phe(4-CN)-OH


Formula: C₁₅H₁₈N₂O₄
MW: 290.319 g/mol
MDL Number: MFCD00672526
CAS Number: 131724-45-3
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -27.0 to -35.0 deg (C=1,DMF)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty