Iguratimod ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

I0945-250MG I0945-25MG
TCI0945-250MGEA 303 USD
TCI0945-250MG TCI0945-25MG
Iguratimod ≥98.0% (by HPLC, titration analysis)
Iguratimod
Formula: C₁₇H₁₄N₂O₆S
MW: 374.37 g/mol
MDL Number: MFCD00882374
CAS Number: 123663-49-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 238.0 to 242.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR