Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(+/-)-Sulpiride ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

S0501-25G S0501-5G
76220-476EA 145 USD InStock
76220-476 76220-478
(+/-)-Sulpiride ≥98.0% (by HPLC, titration analysis)
(+/-)-Sulpiride


Formula: C₁₅H₂₃N₃O₄S
MDL Number: MFCD00055061
CAS Number: 15676-16-1
EINECS: 239-753-7
Merck Index: 14,08989
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 179.0 to 185.0 deg-C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty