Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1,2,3,6,7,8-hexahydropyrene ≥96.0% (by GC)

Supplier: Decon Laboratories Ltd

B5129-1G B5129-5G
TCB5129-1GEA 405 USD InStock
TCB5129-1G TCB5129-5G
4-Bromo-1,2,3,6,7,8-hexahydropyrene ≥96.0% (by GC)
4-Bromo-1,2,3,6,7,8-hexahydropyrene


Formula: C₁₆H₁₅Br
Storage Temperature: Refrigerator
MDL Number: MFCD01571021
CAS Number: 1732-25-8
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 127.0 to 131.0 deg-C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty