Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methylimidazole trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd
Warning

M3214-25G M3214-5G
TCM3214-25GEA 361.79 USD InStock
TCM3214-25G TCM3214-5G
1-Methylimidazole trifluoromethanesulfonate ≥98.0% (by HPLC, titration analysis)
1-Methylimidazole trifluoromethanesulfonate


Formula: C₅H₇F₃N₂O₃S
Melting Pt: 88°C
MDL Number: MFCD18251289
CAS Number: 99257-94-0
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty