Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoro-1-isopropylbenzene 95%

Supplier: AMBEED, INC
Warning

A991032-100MG A991032-250MG
77623-396EA 236.9 USD InStock
77623-396 77623-398
4-Bromo-2-fluoro-1-isopropylbenzene 95%
4-Bromo-2-fluoro-1-isopropylbenzene
Precaución: for Research Use Only


Formula: C₉H₁₀BrF
MW: 217.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20482704
CAS Number: 112611-87-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty