Ethyl 2-oxo-2-(4-(trimethylsilyl)phenyl)acetate ≥97%
Supplier: AOB CHEM USA
|
Warning
|
147516-5G
147516-25G
147516-1G
147516-500MG
77631-330EA
1839.58
USD
77631-330
77635-458
77636-664
77648-700
Ethyl 2-oxo-2-(4-(trimethylsilyl)phenyl)acetate ≥97%
Ethyl 2-oxo-2-(4-(trimethylsilyl)phenyl)acetate
Precaución: For Research Use Only
Formula:
250.366 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34839419 CAS Number: 110193-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...