Rack 'Series 20'
Supplier: SARSTEDT INC
93.841.100 93.844.100 93.846.100 93.848.100 93.849.100 93.893.100
77589-650CS 1066.52 USD
77589-650 77589-652 77589-654 77589-656 77589-658 77589-788
Rack 'Series 20'
Racks Tube Racks
High-quality, safe, and robust racks and boxes are essential for specimen processing and storage. SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Format: 10×2 configuration
  • Space for 20 collection tubes
  • Shatter-proof polycarbonate material ensures that samples are stored securely
  • Special knurling allows SARSTEDT screw cap micro tubes to be opened and closed with one hand
  • Can be autoclaved at 121 °C
  • For storage down to –86 °C
SARSTEDT's practical and versatile range of racks and storage boxes provides a solution for every application.