Block Rack D17
Supplier: SARSTEDT INC
93.852.170 93.852.171 93.852.172 93.852.173 93.852.174 93.852.175 93.851 93.851.171 93.851.172 93.851.173 93.851.174 93.851.175 93.852 93.852.168 93.852.169
77589-682CS 689.45 USD InStock
77589-682 77589-684 77589-686 77589-688 77589-690 77589-692 77589-664 77589-666 77589-668 77589-670 77589-672 77589-674 77589-676 77589-678 77589-680
Block Rack D17
Racks Tube Racks
High-quality, safe, and robust racks and boxes are essential for specimen processing and storage. SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Format: 5×10 configuration
  • 17mm opening diameter
  • Can be autoclaved at 121 °C
  • Available with or without handle
  • Range of colors for each area of the laboratory
SARSTEDT's practical and versatile range of racks and storage boxes provides a solution for every application.
The Block Rack D17 is designed for space-saving sample storage. This rack features 50 apertures for S-Monovettes and tubes with 8 to 17mm diameter.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty