Tube Racks, Series 18
Supplier: SARSTEDT INC
93.870 93.827.013 93.850 93.894 93.826 93.827 93.850.001
77589-730CS 2738.8 USD
77589-730 77589-642 77589-660 77589-790 77589-638 77589-640 77589-662
Tube Racks, Series 18
Racks Tube Racks
High-quality, safe, and robust racks and boxes are essential for specimen processing and storage. SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Format: 6×3 configuration
  • Space for 18 collection tubes
  • Shatter-proof polycarbonate material ensures that samples are stored securely
  • Special knurling allows SARSTEDT screw cap micro tubes to be opened and closed with one hand
  • Can be autoclaved at 121 °C
  • For storage down to –86 °C
SARSTEDT's practical and versatile range of racks and storage boxes provides a solution for every application.