Tube Racks, Series 48
Supplier: SARSTEDT INC
93.868 93.837 93.838 93.838.013 93.048.011 93.1428 93.1431 93.1431.001
77589-728CS 6755.42 USD
77589-728 77589-644 77589-646 77589-648 77589-606 77589-610 77589-612 77589-614
Tube Racks, Series 48
Racks Tube Racks
High-quality, safe, and robust racks and boxes are essential for specimen processing and storage. SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Format: 12×4 configuration
  • Space for 48 collection tubes
  • Shatter-proof polycarbonate material ensures that samples are stored securely
  • Special knurling allows SARSTEDT screw cap micro tubes to be opened and closed with one hand
  • Can be autoclaved at 121 °C
  • For storage down to –86 °C
SARSTEDT's practical and versatile range of racks and storage boxes provides a solution for every application.