2,3-Difluoro-4-(hydroxymethyl)phenol ≥95%

Supplier: AOB CHEM USA
Warning

49967-5G 49967-1G 49967-500MG 49967-25G
77635-116EA 412 USD
77635-116 77639-448 77652-974 77659-862
2,3-Difluoro-4-(hydroxymethyl)phenol ≥95%
2,3-Difluoro-4-(hydroxymethyl)phenol

Precaución: For Research Use Only

Formula: 160.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28789054
CAS Number: 1472068-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR