Acetonitrile 99,8

Supplier: ALADDIN SCIENTIFIC
Danger

A119012-100ML A119012-500ML
ALA119012100MLEA 63.56 USD
ALA119012100ML ALA119012500ML
Acetonitrile 99,8
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Flash Pt: 2 °C
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
Merck Index: 13,00071

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR