tert-Butyl-4-aminopiperidine-1-carboxylate hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC

A121576-5G A121576-1G A121576-250MG A121576-500MG
ALA121576-5GEA 739.01 USD
ALA121576-5G ALA121576-1G ALA121576250MG ALA121576500MG
tert-Butyl-4-aminopiperidine-1-carboxylate hydrochloride ≥97%
tert-Butyl-4-aminopiperidine-1-carboxylate hydrochloride
Formula: C₁₀H₂₁ClN₂O₂
MW: 236.7400 g/mol
MDL Number: MFCD01321014
CAS Number: 189819-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR