Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Azelastine hydrochloride ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A129214-1G A129214-250MG A129214-5G
ALA129214-1GEA 113.91 USD InStock
ALA129214-1G ALA129214250MG ALA129214-5G
Azelastine hydrochloride ≥98% (by HPLC)
Azelastine hydrochloride
Azelastine HCl is a potent, second-generation, selective, histamine antagonistAn H1-receptor antagonist with antiallergic effects and disrupts TNF alpha activity.


Formula: C₂₂H₂₄ClN₃O
Melting Pt: 226 °C
Storage Temperature: Freezer
MDL Number: MFCD00242783
CAS Number: 79307-93-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty