4'-Aminobenzanilide 95
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A132026-1G
A132026-25G
A132026-5G
ALA132026-1GEA
13.21
USD
ALA132026-1G
ALA132026-25G
ALA132026-5G
4'-Aminobenzanilide 95
4'-Aminobenzanilide
Application:4'-Aminobenzanilide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field. It has anticonvulsant property.
Formula:
C₁₃H₁₂N₂O MW: 212.25 g/mol Melting Pt: 127-131°C |
MDL Number:
MFCD00035777 CAS Number: 17625-83-1 EINECS: 241-603-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...