4-Amino-1H-imidazole-5-carbonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A151398-10G A151398-1G A151398-25G
ALA151398-10GEA 354.26 USD
ALA151398-10G ALA151398-1G ALA151398-25G
4-Amino-1H-imidazole-5-carbonitrile ≥98%
4-Amino-1H-imidazole-5-carbonitrile
Formula: C₄H₄N₄
MW: 108.10 g/mol
Melting Pt: 131 °C
Storage Temperature: Ambient
MDL Number: MFCD00129663
CAS Number: 5098-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR