2-Amino-3-chloro-1,4-naphthoquinone ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A151771-25G
A151771-5G
ALA151771-25GEA
57.86
USD
ALA151771-25G
ALA151771-5G
2-Amino-3-chloro-1,4-naphthoquinone ≥98% (by HPLC)
2-Amino-3-chloro-1,4-naphthoquinone
Product Application:Quinoclamine may be used as an analytical reference standard for the determination of the analyte in:Honeybees by dispersive solid-phase extraction (dSPE), followed by liquid and gas chromatography coupled with tandem mass spectrometry (LC-MS/MS and GC-MS/MS).Chlorophyll-containing matrix by dSPE and GC-MS/MS.
Formula:
C₁₀H₆ClNO₂ MW: 207.61 g/mol Melting Pt: 199-205°C |
MDL Number:
MFCD00001680 CAS Number: 2797-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...