2-Amino-2-(4-bromophenyl)acetic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A186179-10G A186179-1G A186179-250MG A186179-5G
ALA186179-10GEA 265.91 USD
ALA186179-10G ALA186179-1G ALA186179250MG ALA186179-5G
2-Amino-2-(4-bromophenyl)acetic acid ≥95%
2-Amino-2-(4-bromophenyl)acetic acid
Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00049327
CAS Number: 71079-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR