Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-TNFRSF11B Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A47765-50UL A47765-100UL
77713-511EA 228.38 USD InStock
77713-511 77713-510
Anti-TNFRSF11B Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:15-1:50

Type:
Antigen: TNFRSF11B
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty