Sulfuric acid 95.0-98.0% ACS
Supplier: VWR Solon Production
|
Danger
|
0499-100ML
0499-500ML
97064-610CS
548.61
USD
97064-610
97064-612
Sulfuric acid 95.0-98.0% ACS
Sulfuric acid
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.84 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8 II |
Specification Test Results
Ammonium | < 0.0002% |
Arsenic | <= 0.01 ppm |
Chloride | <= 0.00002% |
Color (APHA) | <= 10 |
Heavy Metals (as Pb) | <= 0.0001% |
Iron | <= 0.00002% |
Mercury | <= 5 ppb |
Nitrate | <= 0.00005% |
Purity | 95% - 98% |
Residue on Ignition | <= 0.0005% |
Substances Reducing Permanganate | <= 0.0002% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...