Acetonitrile, anhydrous 99,8
Supplier: Origen Biomedical, Inc
|
Danger
|
042311-AK
042311-K2
042311-K7
AA42311-AKEA
75.84
USD
AA42311-AK
AAA42311-K2
AAA42311-K7
Acetonitrile, anhydrous 99,8
Acetonitrile
Anhydrous
Purity: 99.8+%
CAS No.: 75-05-8
Molecular Formula: CH3CN
Formula Weight: 41.05
Flash Point: 5°(41°F)
Physical Form: Liquid
Density: 0.786
Refractive Index: 1.3440
Odor: Pungent
Melting Point: -48 to -45°
Boiling Point: 80-82°
Beilstein Reference: 741857
Merck Reference: 14,70
UN No.: 1648
MDL No.: MFCD00001878
Formula:
H₃CCN MW: 41.05 g/mol Boiling Pt: 81.6 Melting Pt: -45.7 Density: 0.786 Flash Pt: 5 °C (41 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3 II REACH: 01-2119471307-38 Merck Index: 14,70 |
|
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...