A11905-AP
A11905-AU
AAAA11905-APEA
56.7
USD
InStock
AAAA11905-AP
AAA11905-AU
4-Ethylmorpholine 98%
4-Ethylmorpholine
MDL: MFCD00006177
Beilstein Registry No.: 102969
Fieser: 1,383
Purity: 98%
CAS No.: 100-74-3
Molecular Formula: C6H13NO
Formula Weight: 115.18
Flash Point: 27°(80°F)
Refractive Index: 1.4413
Melting Point: -63°
Boiling Point: 138-139°
Beilstein Reference: 102969
UN No.: 2734
MDL No.: MFCD00006177